missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Indinavir sulfate, MedChemExpress
Description
Indinavir sulfate (MK-639) is an orally active and selective HIV-1 protease inhibitor with a Ki of 0.54 nM for PR. Indinavir sulfate exhibits anticancer activity by inhibiting the activation of MMPs-2 hydrolysis, anti-angiogenesis and inducing apoptosis. Indinavir sulfate is also a SARS-CoV 3CLpro inhibitor.
Specifications
Specifications
| Chemical Name or Material | Indinavir sulfate |
| CAS | 157810-81-6 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C36H49N5O8S |
| Quantity | 50 mg |
| Synonym | MK-639 L735524 |
| Solubility Information | DMSO : ≥ 100 mg/mL (140.48 mM) ∣H2O : 50 mg/mL (70.24 mM; Need ultrasonic) |
| SMILES | O=C([C@@H](C[C@H](O)CN(CCN(CC1=CN=CC=C1)C2)[C@@H]2C(NC(C)(C)C)=O)CC3=CC=CC=C3)N[C@H]4C(C=CC=C5)=C5C[C@H]4O.O=S(O)(O)=O |
| Molecular Weight (g/mol) | 711.87 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?