missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Indiko™ and Konelab™ System Glucose Reagents

Product Code. 17857763 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
8 x 20 mL
Unit Size:
Pack of 8
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17857763 8 x 20 mL Pack of 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17857763 Supplier Thermo Scientific™ Supplier No. 981779
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Fully standardize performance with Thermo Scientific™ Indiko™ and Konelab™ System Glucose Reagents, which offer best-in-class methodologies.

Enhance your laboratory's performance with Thermo Scientific™ Indiko™ and Konelab™ System Clinical Chemistry Reagents. These ready-to-use assays come fully validated and compliant, featuring automatic barcoding for efficient process and reordering checks. System reagents provide timely and accurate information, helping to ensure results you can stand behind.

Indiko™ and Konelab™ applications utilize carefully selected methods according to IFCC and other widely used recommendations. Clinical Chemistry methods are provided with system specified multi-calibrators and controls with traceability to international reference materials.

Ordering Information: Not available in the U.S.A., except Glucose Hexokinase.

The Clinical Chemistry Applications for Indiko and Konelab Analyzers cover:

  • Acid Phosphatase
  • Albumin
  • ALP
  • ALT
  • Amylase
  • AST
  • Bilirubin direct and total
  • Calcium
  • Cholesterol
  • HDL and LDL Cholesterol
  • Cholinesterase
  • CO2
  • Creatine Kinase
  • CK-MB
  • Creatinine
  • Gamma-GT
  • Glucose
  • alpha-HBDH
  • Iron
  • LDH
  • Lipase
  • Magnesium
  • Phosphorus
  • Total Protein
  • Triglycerides
  • U/CSF Protein
  • Urea
  • Uric Acid
TRUSTED_SUSTAINABILITY

Specifications

Description Glucose; Kit Size: 8 x 20mL
Detectable Analytes Glucose
Sample Type Plasma, Serum
Type Reagent
Quantity 8 x 20 mL
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.