missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Indiko™ System Clinical Chemistry Reagents, GGT to Uric Acid

Product Code. 17867763 Shop All Thermo Scientific Products
Change view
Click to view available options
Detectable Analytes:
LDH
Triglycerides
Quantity:
4 x 25 mL
12 x 20 mL
Unit Size:
Pack of 12
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Detectable Analytes Quantity Unit Size
17867763 Triglycerides 12 x 20 mL Pack of 12
17848027 LDH 4 x 25 mL -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17867763 Supplier Thermo Scientific™ Supplier No. 981786
Spotlight Offers
Use promo code 28038
View all promotions
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Fully standardize performance with Thermo Scientific™ Indiko™ System Clinical Chemistry Reagents.

Enhance your laboratory's performance with Thermo Scientific™ Indiko™ System Clinical Chemistry Reagents. These ready-to-use assays come fully validated and compliant, featuring automatic barcoding for efficient process and reordering checks. System reagents provide timely and accurate information, helping to ensure results you can stand behind.

  • Indiko™ and Konelab™ applications utilize carefully selected methods according to IFCC and other widely used recommendations
  • Clinical chemistry methods are provided with system specified multi-calibrators and -controls with traceability to international reference materials
  • CE marked kits are compliant with the IVDD
Ordering Information: Not available in the U.S.A.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE Marked
Description Triglycerides; Kit Size: 12 x 20mL
CE Marker Yes
Detectable Analytes Triglycerides
Sample Type Serum
Type Reagent
Quantity 12 x 20 mL
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.