missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Indigokarmine Solutions for PANPEHA indicator paper, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
2.5 L
Unit Size:
2.50 L
Specifications
Specifications
| Chemical Name or Material | Indigokarmine sol |
| Melting Point | -98°C (Data refer to the solvent) |
| Boiling Point | 64°C to 65°C (1013 hPa) |
| UN Number | UN1230 |
| Quantity | 2.5 L |
Safety and Handling
Product Identifier
- Indigokarminlösung für PANPEHA-Indikatorpapier
- Danger
- Acute toxicity Category 3
- Flammable liquid Category 2
- Specific target organ toxicity after single exposure Category 1
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P243-Take precautionary measures against static discharge.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : methanol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction