missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Indigo (Technical Grade), TRC
Specifications
Specifications
| Quantity | 10 g |
| Synonym | 3H-Indol-3-one, 2-(1,3-dihydro-3-oxo-2H-indol-2-ylidene)-1,2-dihydro-, Indigo Pure BASF (6CI), [δ2,2'-Biindoline]-3,3'-dione (8CI), 11669 Blue, 2,2'-Bis(2,3-dihydro-3-oxoindolylidene), 2-(1,3-Dihydro-3-oxo-2H-indol-2-ylidene)-1,2-dihydro-3H-indol-3-one, Blue No. 201, C Blue 22, C.I. 73000, C.I. Natural Blue 1, C.I. Pigment Blue 66, C.I. Vat Blue 1, Cystoceva, D and C Blue No. 6, D&C Blue No. 6, D+C Blue No. 6, Diindogen, Dystar Indigo Gr, Indigo, Indigo Blue, Indigo Ciba, Indigo Ciba SL, Indigo J, Indigo N, Indigo NAC, Indigo NACCO, Indigo P, Indigo PLN, Indigo Powder W, Indigo Pure BASF Powder K, Indigo Synthetic, Indigo VS, Indigotin, Indigotin (natural), Indigotine, Japan Blue 201, Lithosol Deep Blue B, Mitsui Indigo Paste, Mitsui Indigo Pure, Mitsui Indigo Pure EXN, Monolite Fast Navy Blue BV, Natural Blue 1, Natural blue indigotin, Pigment Blue 66, Pigment Indigo, Pigment Indigo V, Reduced Dark Blue VB, Synthetic Indigo TS, Synthetic indigo, Vat Blue 1, Vat Dark Blue VB, Vulcafix Blue R, Vulcafor Blue A, Vulcanosine Dark Blue L, Vulcol Fast Blue GL, Vynamon Blue A, [δ2,2'(3H,3'H)-Biindole]-3,3'-dione, δ2,2'-Bipseudoindoxyl |
| InChI Formula | InChI=1S/C16H10N2O2/c19-15-9-5-1-3-7-11(9)17-13(15)14-16(20)10-6-2-4-8-12(10)18-14/h1-8,17-18H/b14-13+ |
| Formula Weight | 262.0742 |
| Chemical Name or Material | Indigo |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?