missing translation for 'onlineSavingsMsg'
Learn More

Indigo Carmine, TRC

Product Code. 30397362
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30397362 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30397362 Supplier Toronto Research Chemicals Supplier No. TRCI5213535G
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Indigotine (E132)
CAS 860-22-0
Molecular Formula C16 H8 N2 O8 S2 . 2 Na
Quantity 5 g
Synonym 1H-Indole-5-sulfonic acid, 2-(1,3-dihydro-3-oxo-5-sulfo-2H-indol-2-ylidene)-2,3-dihydro-3-oxo-, sodium salt (1:2), 1H-Indole-5-sulfonic acid, 2-(1,3-dihydro-3-oxo-5-sulfo-2H-indol-2-ylidene)-2,3-dihydro-3-oxo-, disodium salt (9CI), C.I. Acid Blue 74 (6CI), [Δ2,2'-Biindoline]-5,5'-disulfonic acid, 3,3'-dioxo-, disodium salt (8CI), 12070 Blue, 1311 Blue, 5,5'-Indigodisulfonic acid disodium salt, A.F. Blue No. 2, Acid Blue 74, Acid Blue W, Acid Leather Blue IC, Airedale Blue IN, Amacid Brilliant Blue, Aniline Carmine Powder, Ariavit Indigo Carmine, Atul Indigo Carmine, Basovit Blue 665E, Blue 2, Blue No. 2, Bucacid Indigotine B, C.I. 73015, C.I. 75781, C.I. Food Blue 1, C.I. Natural Blue 2, Canacert Indigo Carmine, Carmine Blue, Carmine indigo, Cilefa Blue R, Cogilor Blue 511.11, Disodium 5,5'-indigodisulfonate, Disodium 5,5'-indigotin disulfonate, Dolkwal Indigo Carmine, E 132, Edicol Supra Blue X, Eurocert Indigo Carmine 311811, FD & C Blue 2, FD & C Blue No 2-307045, FD and C Blue 2, FD and C Blue No. 2, FD&C Blue No. 2, FD&C Blue No. 2-37006, Food Blue 1, Food Blue 2, Food Blue No. 1, Food Blue No. 2, Grape Blue A, HD Indigo Carmine, HD Indigo Carmine Supra, Hexacert Blue No. 2, Hexacol Indigo Carmine Supra, Indigo Carmine 307019, Indigo Carmine 36009, Indigo Carmine 37006, Indigo Carmine A, Indigo Carmine AC, Indigo Carmine BP, Indigo Carmine Conc. FQ, Indigo Carmine Powder, Indigo Carmine X, Indigo Extract, Indigo carmine, Indigo carmine 85E132, Indigo carmine E 132, Indigo carmine NB, Indigotin, Indigotin (solubilized), Indigotine, Indigotine B, Indigotine Blue LZ, Indigotine Carmine, Indigotine Extra Pure A, Indigotine I, Indigotine IA, Indigotine Lake, Indigotine N, Indigotine disodium salt, Indocarmine F, Intense Blue, Japan Blue 2, Japan Food Blue No. 2, L Blue Z 5010, Maple Indigo Carmine, Mitsui Indigo Carmine, Necol Indigo Carmine, Neelicol Indigo Carmine, San-ei Indigo Carmine, Sepisperse Dry 1003, Sodium 5,5'-indigodisulfonate, Sodium 5,5'-indigotindisulfonate, Soluble indigo, Soluble indigo blue, Sumitomo Wool Blue SBC, Usacert Blue No. 2, Usacert FD & C Blue No 2-310118, Usacert FD and C Blue No. 2, WAS 35, Water Blue 177557, Water Blue 177558
InChI Formula InChI=1S/C16H10N2O8S2.2Na/c19-15-9-5-7(27(21,22)23)1-3-11(9)17-13(15)14-16(20)10-6-8(28(24,25)26)2-4-12(10)18-14;;/h1-6,17-18H,(H,21,22,23)(H,24,25,26);;/q;2*+1/p-2/b14-13+;;
SMILES [Na+].[Na+].[O-]S(=O)(=O)c1ccc2N\C(=C/3\Nc4ccc(cc4C3=O)S(=O)(=O)[O-])\C(=O)c2c1
IUPAC Name disodium;(2E)-3-oxo-2-(3-oxo-5-sulfonato-1H-indol-2-ylidene)-1H-indole-5-sulfonate
Molecular Weight (g/mol) 466.35
Formula Weight 465.9517
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.