missing translation for 'onlineSavingsMsg'
Learn More

Indigo Carmine, MP Biomedicals™

Product Code. 11457982 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
Unit Size:
100g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11457982 100 g 100g
11447982 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11457982 Supplier MP Biomedicals Supplier No. 0215263880
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Indigo Carmine is used as a stain.

  • Indigo Carmine is useful in the staining of Negri bodies.
  • It can be used as a contrast stain for plasma when red nuclear stains are used.

Chemical Identifiers

CAS 860-22-0
Molecular Formula C16H8N2Na2O8S2
Molecular Weight (g/mol) 466.35
MDL Number MFCD00005723
InChI Key KHLVKKOJDHCJMG-QDBORUFSSA-L
Synonym Acid Blue 74, Indigo-5, 5'-disulfonic acid disodium salt
PubChem CID 5284351
IUPAC Name disodium (E)-3,3'-dioxo-1H,1'H,3H,3'H-[2,2'-biindolylidene]-5,5'-disulfonate
SMILES [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2N\C(C(=O)C2=C1)=C1\NC2=CC=C(C=C2C1=O)S([O-])(=O)=O

Specifications

Absorbance λ max (water) 608 to 610 nm
Assay Percent Range ˜85%
Color Purple
Solubility Information 10 to 50 mg/mL at 70.7°F
Quantity 100 g
Physical Form Powder
Formula Weight 466.3
Grade Histological
Chemical Name or Material Indigo carmine, Cell Biology and Analysis, Molecular Biology

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.