missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Indigo Carmine, for analysis, high purity biological stain

Product Code. 10724471 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10724471 25 g Glass bottle 25g
10215680 100 g Glass bottle 100g
10734471 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10724471 Supplier Thermo Scientific Chemicals Supplier No. 157440250
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 860-22-0 | C16H8N2Na2O8S2 | 466.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 860-22-0
Molecular Formula C16H8N2Na2O8S2
Molecular Weight (g/mol) 466.35
MDL Number MFCD00005723
InChI Key KHLVKKOJDHCJMG-QDBORUFSSA-L
Synonym Acid Blue 74, C.I. 73015, 5, 5'-Indigodisulfonic acid, disodium salt
PubChem CID 5284351
IUPAC Name disodium;(2E)-3-oxo-2-(3-oxo-5-sulfonato-1H-indol-2-ylidene)-1H-indole-5-sulfonate
SMILES [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2N\C(C(=O)C2=C1)=C1\NC2=CC=C(C=C2C1=O)S([O-])(=O)=O

Specifications

Dye Content 85% min.
Infrared Spectrum Authentic
Packaging Glass bottle
Beilstein 25, 304
Merck Index 15, 4982
Color Blue
Solubility Information Solubility in water: 1g/100mL (25 c). Other solubilities: slightly soluble in alcohol, practically insoluble in most other organic, solvents
Quantity 25 g
Physical Form Fine Crystalline Powder
Formula Weight 466.34
Grade Analysis
Chemical Name or Material Indigo Carmine
Show More Show Less
compliance-icons
Product Identifier
  • Indigo Carmine
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.