Learn More
Indian Hedgehog/Ihh Antibody (S03-3F2), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-19878-100ul
 View more versions of this product
Product Code. 18678103
  • £500.00 / 100µL
Description
Indian Hedgehog/Ihh Monoclonal antibody specifically detects Indian Hedgehog/Ihh in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Indian Hedgehog/Ihh
Monoclonal
0.3 mg/mL
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Rabbit
Affinity purified
RUO
Primary
Human, Mouse, Rat
Purified
Western Blot
S03-3F2
Unconjugated
BDA1Indian hedgehog homolog (Drosophila), HHG2, HHG-2, Indian hedgehog, Indian hedgehog (Drosophila) homolog, Indian hedgehog homolog, indian hedgehog protein
A synthetic peptide of human Indian Hedgehog/Ihh (Uniprot # Q14623)
100 μg
Stem Cell Signaling Pathway
3549
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.