missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Indian Hedgehog/Ihh Antibody (S03-3F2), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19878-50ul
This item is not returnable.
View return policy
Description
Indian Hedgehog/Ihh Monoclonal antibody specifically detects Indian Hedgehog/Ihh in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Indian Hedgehog/Ihh | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| S03-3F2 | |
| Unconjugated | |
| BDA1Indian hedgehog homolog (Drosophila), HHG2, HHG-2, Indian hedgehog, Indian hedgehog (Drosophila) homolog, Indian hedgehog homolog, indian hedgehog protein | |
| A synthetic peptide of human Indian Hedgehog/Ihh (Uniprot # Q14623) | |
| 50 μg | |
| Stem Cell Signaling Pathway | |
| 3549 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction