missing translation for 'onlineSavingsMsg'
Learn More

Indian Hedgehog/Ihh Antibody (9L16), Allophycocyanin, Novus Biologicals™

Product Code. 18470442 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18470442 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18470442 Supplier Novus Biologicals Supplier No. NBP222020APC

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

Indian Hedgehog/Ihh Monoclonal antibody specifically detects Indian Hedgehog/Ihh in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Indian Hedgehog/Ihh
Applications Western Blot, ELISA
Classification Monoclonal
Clone 9L16
Conjugate APC
Dilution Western Blot, ELISA
Formulation PBS
Gene Alias BDA1Indian hedgehog homolog (Drosophila), HHG2, HHG-2, Indian hedgehog, Indian hedgehog (Drosophila) homolog, Indian hedgehog homolog, indian hedgehog protein
Host Species Rat
Immunogen Human recombinant Indian Hedgehog/Ihh (N-terminus)
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 3549
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.