missing translation for 'onlineSavingsMsg'
Learn More

Indeglitazar, MedChemExpress

Product Code. 16486188
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16486188 10 mg 10mg
16448787 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16486188 Supplier Medchem Express Supplier No. HY14817
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Indeglitazar (PPM 204) is an orally available PPAR pan-agonist for all three PPARα, PPARδ and PPARγ.

Indeglitazar (PPM 204) is an orally available PPAR pan-agonist for all three PPARα, PPARδ and PPARγ.

Specifications

Chemical Name or Material Indeglitazar
CAS 835619-41-5
Color Gray
Purity Grade Notes Research
Molecular Formula C19H19NO6S
Quantity 10 mg
Synonym PPM 204
Solubility Information DMSO : 100 mg/mL (256.79 mM; Need ultrasonic)
SMILES O=C(O)CCC1=CN(S(=O)(C2=CC=C(OC)C=C2)=O)C3=C1C=C(OC)C=C3
Molecular Weight (g/mol) 389.42
Formula Weight 389.42
Percent Purity 99.11%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.