missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Indapamide, MedChemExpress
Description
Indapamide is an orally active sulphonamide diuretic agent, that can reduce blood pressure by decreasing vascular reactivity and peripheral vascular resistance. Indapamide is also can reduce left ventricular hypertrophy.
Specifications
Specifications
| Chemical Name or Material | Indapamide |
| CAS | 26807-65-8 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C16H16ClN3O3S |
| Quantity | 1 g |
| Solubility Information | DMSO : ≥ 100 mg/mL (273.35 mM) ∣H2O : 0.67 mg/mL (1.83 mM; Need ultrasonic) |
| SMILES | O=C(NN1C(C)CC2=C1C=CC=C2)C3=CC=C(Cl)C(S(=O)(N)=O)=C3 |
| Molecular Weight (g/mol) | 365.83 |
| Formula Weight | 365.83 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?