missing translation for 'onlineSavingsMsg'
Learn More

Indane, TRC

Product Code. 30392768
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
Product Code. Quantity unitSize
30392768 5 g 5g
1 options
This item is not returnable. View return policy

Product Code. 30392768

Brand: Toronto Research Chemicals TRCI5080005G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Indan
CAS 496-11-7
Molecular Formula C9 H10
Quantity 5 g
Synonym 1H-Indene, 2,3-dihydro-, Indan (8CI), 2,3-Dihydro-1H-indene, 1,2-Hydrindene, 2,3-Dihydroindene, Benzocyclopentane, Hydrindene, Hydrindonaphthene, Indane, NSC 5292
InChI Formula InChI=1S/C9H10/c1-2-5-9-7-3-6-8(9)4-1/h1-2,4-5H,3,6-7H2
SMILES C1Cc2ccccc2C1
IUPAC Name 2,3-dihydro-1H-indene
Molecular Weight (g/mol) 118.176
Formula Weight 118.078
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.