missing translation for 'onlineSavingsMsg'
Learn More

Indane, 95%

Product Code. 10349060 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10349060 100 g Glass bottle 100g
10410531 5 g Glass bottle 5g
10073460 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10349060 Supplier Thermo Scientific Acros Supplier No. 292481000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 496-11-7 | C9H10 | 118.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 496-11-7
MDL Number MFCD00003795
InChI Key PQNFLJBBNBOBRQ-UHFFFAOYSA-N
Synonym indan, indane, hydrindene, benzocyclopentane, 1h-indene, 2,3-dihydro, 2,3-dihydroindene, 1,2-hydrindene, hydrindonaphthene, indane alkane, indene, 2,3-dihydro
PubChem CID 10326
ChEBI CHEBI:37911
IUPAC Name 2,3-dihydro-1H-indene
SMILES C1CC2=CC=CC=C2C1

Specifications

Melting Point -51°C
Color Colorless to Yellow
Density 0.9600g/mL
Boiling Point 176°C
Flash Point 50°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Molecular Formula C9H10
Refractive Index 1.5350 to 1.538
Quantity 100 g
Beilstein 05,II,376
Merck Index 15, 4971
Specific Gravity 0.96
Solubility Information Solubility in water: insoluble. Other solubilities: misciple in ethanol,diethyl ether;,slightly soluble in chloroform
Molecular Weight (g/mol) 118.18
Formula Weight 118.18
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Indane, 95%
Show More Show Less
compliance-icons
Product Identifier
  • Indan
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.