missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Indan-1-one (1-Indanone), Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
Indan-1-one (1-Indanone)
Specifications
Specifications
| Chemical Name or Material | 1-Indanone |
| CAS | 83-33-0 |
| Analyte or Component Names | 1-Indanone |
| Shipping Condition | Room Temperature |
| Molecular Formula | C9 H8 O |
| Certifications/Compliance | ISO 9001 |
| Synonym | 1H-Inden-1-one, 2,3-dihydro-, 1-Indanone (8CI), Indanone (7CI), 2,3-Dihydro-1H-inden-1-one, 1-Indone, 1-Oxobenzocyclopentane, 1H-Indan-1-one, 2,3-Dihydro-1-indenone, NSC 2581, α-Hydrindone, α-Indanone |
| InChI Formula | InChI=1S/C9H8O/c10-9-6-5-7-3-1-2-4-8(7)9/h1-4H,5-6H2 |
| SMILES | O=C1CCc2ccccc12 |
| IUPAC Name | 2,3-dihydroinden-1-one |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction