missing translation for 'onlineSavingsMsg'
Learn More

Indacrinone, MedChemExpress

Product Code. 16422478
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16422478 5 mg 5mg
16410279 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16422478 Supplier Medchem Express Supplier No. HYU00242
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Indacrinone (MK196) is an investigational diuretic which has pronounced saluretic activity in the rat and dog as well as both uricosuric and saluretic activity in the chimpanzee.

Indacrinone (MK196) is an investigational diuretic which has pronounced saluretic activity in the rat and dog as well as both uricosuric and saluretic activity in the chimpanzee.

Specifications

Chemical Name or Material Indacrinone
CAS 56049-88-8
Purity Grade Notes Research
Molecular Formula C18H14Cl2O4
Quantity 5 mg
Synonym MK-196 Indacrynic acid
SMILES O=C(O)COC1=CC2=C(C(C(C3=CC=CC=C3)(C)C2)=O)C(Cl)=C1Cl
Molecular Weight (g/mol) 365.21
Formula Weight 365.21
Grade Research
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.