missing translation for 'onlineSavingsMsg'
Learn More

Indacaterol Maleic Acid Salt, TRC

Product Code. 17938324
Change view
Click to view available options
Quantity:
1 mg
5 mg
25 mg
Unit Size:
1mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17938324 25 mg 25mg
17908124 5 mg 5mg
17968594 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17938324 Supplier Toronto Research Chemicals Supplier No. TRCI49975025MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 753498-25-8
Molecular Formula C24 H28 N2 O3 . C4 H4 O4
Molecular Weight (g/mol) 508.56
Synonym 2(1H)-Quinolinone, 5-[(1R)-2-[(5,6-diethyl-2,3-dihydro-1H-inden-2-yl)amino]-1-hydroxyethyl]-8-hydroxy-, (2Z)-2-butenedioate (1:1), 2(1H)-Quinolinone, 5-[(1R)-2-[(5,6-diethyl-2,3-dihydro-1H-inden-2-yl)amino]-1-hydroxyethyl]-8-hydroxy-, (2Z)-2-butenedioate (1:1) (salt) (9CI), Arcapta, Indacaterol maleate, Onbrez, QAB 149AFA
IUPAC Name (Z)-but-2-enedioic acid;5-[(1R)-2-[(5,6-diethyl-2,3-dihydro-1H-inden-2-yl)amino]-1-hydroxyethyl]-8-hydroxy-1H-quinolin-2-one
SMILES CCc1cc2CC(Cc2cc1CC)NC[C@H](O)c3ccc(O)c4NC(=O)C=Cc34.OC(=O)\C=C/C(=O)O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C24H28N2O3.C4H4O4/c1-3-14-9-16-11-18(12-17(16)10-15(14)4-2)25-13-22(28)19-5-7-21(27)24-20(19)6-8-23(29)26-24;5-3(6)1-2-4(7)8/h5-10,18,22,25,27-28H,3-4,11-13H2,1-2H3,(H,26,29);1-2H,(H,5,6)(H,7,8)/b;2-1-/t22-;/m0./s1
Formula Weight 508.221
Chemical Name or Material Indacaterol Maleic Acid Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.