missing translation for 'onlineSavingsMsg'
Learn More

Indacaterol Maleate, Mikromol

Product Code. 30458343
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30458343 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30458343 Supplier Mikromol Supplier No. MM3994.000250
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Indacaterol Maleate, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Indacaterol Maleic Acid Salt
CAS 753498-25-8
Analyte or Component Names Indacaterol Maleic Acid Salt
Shipping Condition Room Temperature
Molecular Formula C24 H28 N2 O3 . C4 H4 O4
Certifications/Compliance ISO 17034
Synonym 2(1H)-Quinolinone, 5-[(1R)-2-[(5,6-diethyl-2,3-dihydro-1H-inden-2-yl)amino]-1-hydroxyethyl]-8-hydroxy-, (2Z)-2-butenedioate (1:1), 2(1H)-Quinolinone, 5-[(1R)-2-[(5,6-diethyl-2,3-dihydro-1H-inden-2-yl)amino]-1-hydroxyethyl]-8-hydroxy-, (2Z)-2-butenedioate (1:1) (salt) (9CI), Arcapta, Indacaterol maleate, Onbrez, QAB 149AFA
InChI Formula InChI=1S/C24H28N2O3.C4H4O4/c1-3-14-9-16-11-18(12-17(16)10-15(14)4-2)25-13-22(28)19-5-7-21(27)24-20(19)6-8-23(29)26-24;5-3(6)1-2-4(7)8/h5-10,18,22,25,27-28H,3-4,11-13H2,1-2H3,(H,26,29);1-2H,(H,5,6)(H,7,8)/b;2-1-/t22-;/m0./s1
Solution Type Neat
SMILES CCc1cc2CC(Cc2cc1CC)NC[C@H](O)c3ccc(O)c4NC(=O)C=Cc34.OC(=O)\C=C/C(=O)O
IUPAC Name (Z)-but-2-enedioic acid;5-[(1R)-2-[(5,6-diethyl-2,3-dihydro-1H-inden-2-yl)amino]-1-hydroxyethyl]-8-hydroxy-1H-quinolin-2-one
Molecular Weight (g/mol) 508.56
Quantity 250 mg
Formula Weight 508.221 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.