missing translation for 'onlineSavingsMsg'
Learn More

Indacaterol maleate, MedChemExpress

Product Code. 16496436
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16496436 100 mg 100mg
16431726 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16496436 Supplier Medchem Express Supplier No. HY14299A
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Indacaterol (QAB149) maleate is an ultra-long-acting β-adrenoceptor agonist.

Indacaterol (QAB149) maleate is an ultra-long-acting β-adrenoceptor agonist.

Specifications

Chemical Name or Material Indacaterol maleate
CAS 753498-25-8
Color White
Purity Grade Notes Research
Molecular Formula C28H32N2O7
Quantity 100 mg
Synonym QAB149
Solubility Information DMSO : 100 mg/mL (196.63 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES O=C1NC2=C(C([C@@H](O)CNC3CC4=C(C=C(CC)C(CC)=C4)C3)=CC=C2O)C=C1.O=C(O)/C=C\C(O)=O
Molecular Weight (g/mol) 508.56
Formula Weight 508.56
Percent Purity 96.59%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.