missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Incubator BBD6220 220L SS + FFO O2-control + UK Pl

Brand:  Thermo Scientific 51020241+51900763+51900303

21717.99 GBP valid until 2026-05-31
BEST PRICE on Promo! Use promo code "27998" to get your promotional price.


Incubator BBD6220 220L SS + FFO O2-control + UK Plug

Product Code. 17408082

  • ONLINE EXCLUSIVE PRICE
    £31545.00
    £28707.75 / Pack of 1
    Save £2837.25
    8% Off
View all promotions

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.