missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel, Ni/Cr/Mo/Nb, Alloy 625, N06625, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Nickel, Ni/Cr/Mo/Nb, Alloy 625, N06625, Ø40x20mm
Specifications
Specifications
| Chemical Name or Material | Phosphorus/Carbon/Boron/Aluminium/Cobalt/Titanium/Tantalum/Copper/Manganese/Silicon/Niobium/Iron/Molybdenum/Chromium/Nickel |
| CAS | 7440-02-0 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Phosphorus 0.002 %/Carbon 0.0026 %/Boron 0.004 %/Aluminium 0.006 %/Cobalt 0.008 %/Titanium 0.0096 %/Tantalum 0.0112 %/Copper 0.057 %/Manganese 0.0694 %/Silicon 0.251 %/Niobium 2.64 %/Iron 4.22 %/Molybdenum 9.6 %/Chromium 20.22 %/Nickel 62.93 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction