missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel, Ni/Cr, Alloy 600, N06600, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Nickel, Ni/Cr, Alloy 600, N06600, Ø40x15mm
Specifications
Specifications
| Chemical Name or Material | Magnesium/Aluminium/Titanium/Copper/Manganese/Cobalt/Iron/Chromium |
| CAS | 7440-02-0 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Magnesium 0.01 %/Aluminium 0.025 %/Titanium 0.22 %/Copper 0.42 %/Manganese 0.47 %/Cobalt 0.62 %/Iron 7.1 %/Chromium 15.56 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction