Learn More
Inconel™ 600 gauze, 60 mesh woven from 0.19mm (0.0075 in.) dia. wire, Thermo Scientific™
Brand: Thermo Scientific Alfa Aesar 046584.HB
Code : NEW
Additional Details : Weight : 0.01000kg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Inconel™ 600 gauze | |
Ni:Cr:Fe; 77:15.5:7.5 wt% | |
300 x 300 mm | |
Wire Cloth |
60 mesh woven from 0.19mm (0.0075 in.) dia. wire | |
MFCD01741927 | |
60 Mesh |