missing translation for 'onlineSavingsMsg'
Learn More
Learn More
INCENP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-62620-25ul
This item is not returnable.
View return policy
Description
INCENP Polyclonal antibody specifically detects INCENP in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| INCENP | |
| Polyclonal | |
| Immunohistochemistry 1:2500 - 1:5000, Immunohistochemistry-Paraffin 1:2500 - 1:5000 | |
| binds and activates aurora-B and -C in vivo and in vitro, chromosomal passenger protein, FLJ31633, inner centromere protein, inner centromere protein antigens (135kD, 155kD), inner centromere protein antigens 135/155kDa, inner centromere protein INCENP, MGC111393 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: KLMEFLCNMDNKDLVWLEEIQEEAERMFTREFSKEPELMPKTPSQKNRRKKRRISYVQDENRDPIRRRLSRRKSRSSQLSSRRLRSKDSVEKLATVVGE | |
| 25 μL | |
| Cell Cycle and Replication, Chromatin Research, DNA Repair, Mitotic Regulators, Protein Kinase | |
| 3619 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction