missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX InCELL Hunter™ U2OS Bcl2-Bax Protein Binding Cell Line
Click to view available options
Quantity:
2 Vials
Unit Size:
2 ampoules
Description
The InCELL Hunter™ U2OS Bcl2-Bax Protein Binding Cell Line provides an easy-to-use cell based assay to study small molecule drug candidates that disrupt protein binding. This kit assesses compound-based disruption of Bcl2-Bax binding via detection of Bax levels. This product uses EFC technology. The cell line included overexpresses Bcl2 target in an untagged form and Bax binding partner fused to PL. When a compound engages the Bcl2 target protein, partner binding is disrupted and the destabilized Bax is degraded, leading to a loss of EFC signal upon addition of EA containing detection reagent. This stable modified cell line is provided as two vials of cryopreserved cells.
Specifications
Specifications
| Assay Entry | Target Engagement |
| Cell Line | U2OS |
| For Use With (Application) | Cell Lines |
| Primary Agonist | ABT-737 |
| Quantity | 2 Vials |
| Target Entry | Bcl2-Bax |
| Accession Number | NM_000633 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction