missing translation for 'onlineSavingsMsg'
Learn More

DiscoverX InCELL Hunter™ Jurkat RIPK3 Kinase Binding Cell Line

Product Code. 17334829
Encompass_Preferred
Click to view available options
Quantity:
2 Vials
Unit Size:
2 ampoules
This item is not returnable. View return policy

Product Code. 17334829

Brand: DiscoverX 960077C19

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The InCELL Hunter™ Jurkat RIPK3 Kinase Binding Cell Line measures target engagement via compound-induced stabilization of RIPK3 target protein. This cell line ships as two vials of cryopreserved cells.

The InCELL Hunter™ Jurkat RIPK3 Kinase Binding Cell Line is a stable, engineered cell line for use in studying small molecule drug candidates that engage with kinases. This cell line enables assessment of compound-engagement-based stabilization of RIPK3 via detection of RIPK3 levels. This product uses EFC technology. The cell line included overexpresses RIPK3 target protein fused to PL. In the absence of compound that binds and stabilizes the target, this protein is denatured by the heat treatment and no EFC signal is produced. When compound engages and heat-stabilizes the target, EA contained in the detection reagent produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells.

Specifications

Assay Entry Intracellular Binding
Cell Line Jurkat
For Use With (Application) Cell Lines
Quantity 2 Vials
Target Entry RIPK3
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.