missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX InCELL Hunter™ A549 GLP Methyltransferase Cell Line
Click to view available options
Quantity:
2 Vials
Unit Size:
2 ampoules
Description
The InCELL Hunter™ A549 GLP Methyltransferase Cell Line is a stable, engineered cell line for use in studying small molecule drug candidates that engage with regulators of epigenetics pathways. This cell line enables assessment of compound-engagement-based stabilization of GLP via detection of GLP levels. This product uses EFC technology. The cell line included overexpresses GLP target protein fused to PL. In the absence of compound that binds and stabilizes the target, this protein is degraded and no EFC signal is produced. When compound engages and stabilizes the target, EA contained in the detection reagent produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells.
Specifications
Specifications
| Assay Entry | Target Engagement |
| Cell Line | A549 |
| For Use With (Application) | Cell Lines |
| Primary Agonist | UNC 0638 |
| Quantity | 2 Vials |
| Target Entry | GLP |
| Accession Number | NM_024757 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction