missing translation for 'onlineSavingsMsg'
Learn More

INCB3344, MedChemExpress

Product Code. 16453997
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16453997

Brand: Medchem Express HY50674

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress INCB3344 is a potent, selective and orally bioavailable CCR2 antagonist with IC50 values of 5.1 nM (hCCR2) and 9.5 nM (mCCR2) in binding antagonism and 3.8 nM (hCCR2) and 7.8 nM (mCCR2) in antagonism of chemotaxis activity.

INCB3344 is a potent, selective and orally bioavailable CCR2 antagonist with IC50 values of 5.1 nM (hCCR2) and 9.5 nM (mCCR2) in binding antagonism and 3.8 nM (hCCR2) and 7.8 nM (mCCR2) in antagonism of chemotaxis activity.

Specifications

Chemical Name or Material INCB3344
CAS 1262238-11-8
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C29H34F3N3O6
Quantity 50 mg
Solubility Information DMSO : 220 mg/mL (380.89 mM; Need ultrasonic)
SMILES CCO[C@H]1CN([C@]2([H])CC[C@@](CC2)(C3=CC4=C(OCO4)C=C3)O)C[C@@H]1NC(CNC(C5=CC(C(F)(F)F)=CC=C5)=O)=O
Molecular Weight (g/mol) 577.59
Formula Weight 577.59
Percent Purity 96.59%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.