missing translation for 'onlineSavingsMsg'
Learn More
Learn More
INCB 3284, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
INCB 3284 is a potent, selective and orally bioavailable human CCR2 antagonist, inhibiting monocyte chemoattractant protein-1 binding to hCCR2, with an IC50 of 3.7 nM. INCB 3284 can be used in the research of acute liver failure.
Specifications
Specifications
| Chemical Name or Material | INCB 3284 |
| CAS | 887401-92-5 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C26H31F3N4O4 |
| Quantity | 10 mg |
| Solubility Information | DMSO : ≥ 83.3 mg/mL (160.03 mM) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble) |
| SMILES | O=C(NCC(N[C@H]1CN([C@H]2CC[C@@](C3=CC=C(OC)N=C3)(O)CC2)CC1)=O)C4=CC=CC(C(F)(F)F)=C4 |
| Molecular Weight (g/mol) | 520.54 |
| Formula Weight | 520.54 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction