missing translation for 'onlineSavingsMsg'
Learn More

Furosemide, MedChemExpress

Product Code. 16400788
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 16400788

Brand: Medchem Express HYB0135

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Furosemide is a potent and orally active inhibitor of Na+/K+/2Cl- (NKCC) cotransporter, NKCC1 and NKCC2. Furosemide is also a GABAA receptors antagonist and displays 100-fold selectivity for α6-containing receptors than α1-containing receptors. Furosemide acts as a loop diuretic and used for the study of congestive heart failure, hypertension and edema.

Furosemide is a potent and orally active inhibitor of Na+/K+/2Cl- (NKCC) cotransporter, NKCC1 and NKCC2. Furosemide is also a GABAA receptors antagonist and displays 100-fold selectivity for α6-containing receptors than α1-containing receptors. Furosemide acts as a loop diuretic and used for the study of congestive heart failure, hypertension and edema.

Specifications

Chemical Name or Material Furosemide
CAS 54-31-9
Color White
Purity Grade Notes Research
Molecular Formula C12H11ClN2O5S
Quantity 5 g
Solubility Information DMSO : ≥ 100 mg/mL (302.35 mM) ∣H2O : 0.1 mg/mL (0.30 mM; Need ultrasonic)
SMILES O=C(O)C1=CC(S(=O)(N)=O)=C(Cl)C=C1NCC2=CC=CO2
Molecular Weight (g/mol) 330.74
Formula Weight 330.74
Percent Purity 99.43%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.