Learn More
INCA1 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 30441-1-AP-150UL
Description
The INCA1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human INCA1. This antibody recognizes human, mouse, rat antigen. The INCA1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| INCA1 | |
| 550 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| HSD45, INCA1, Protein INCA1 | |
| 150 μL | |
| 388324 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |