missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Inavolisib, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Description
GDC-0077 (RG6114) is a potent, orally available, and selective PI3Kα inhibitor (IC50=0.038 nM). GDC-0077 (RG6114) exerts its activity by binding to the ATP binding site of PI3K, thereby inhibiting the phosphorylation of PIP2 to PIP3. GDC-0077 (RG6114) is more selective for mutant versus wild-type PI3Kα.
Specifications
Specifications
| Chemical Name or Material | Inavolisib |
| CAS | 2060571-02-8 |
| Color | Tan |
| Purity Grade Notes | Research |
| Molecular Formula | C18H19F2N5O4 |
| Quantity | 5 mg |
| Synonym | GDC-0077 RG6114 |
| Solubility Information | DMSO : 110 mg/mL (270.02 mM; Need ultrasonic) |
| SMILES | C[C@@H](C(N)=O)NC1=CC=C(C2=NC(N3[C@H](C(F)F)COC3=O)=CN2CCO4)C4=C1 |
| Molecular Weight (g/mol) | 407.37 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction