Learn More
Ohaus™ In-Use Cover, R71
Protects your compact scale from dirt, dust, spills, and fingerprints during use.
Brand: Ohaus™ 30135320
Code : NEW
Additional Details : Weight : 0.50000kg
Description
- Designed to fit R71 series compact scales:
- R71MD3 AM
- R71MD6 AM
- R71MD15 AM
- R71MD35 AM
- R71MHD3 AM
- R71MHD6 AM
- R71MHD15 AM
- R71MHD35 AM
- R71MD60 AM
- Protects your compact scale from dust, dirt, spills, and fingerprints
Specifications
R71 Series Compact Scale | |
Chinese, English (US), French, German, Italian, Japanese, Korean, Polish, Portuguese (BR), Russian, Spanish (MX) | |
1/Pk. | |
In-Use Covers |