Learn More
Mettler Toledo™ In-Use Balance Covers
Product Code. 12750734 Shop All Mettler Toledo Products
Change view
Click to view available options
For Use With (Equipment):
NewClassic MS-S 0.01 /0.1 g Balance
PHS, NewClassic MS 0.1/1 mg Balance
Weighing Platform “S”, XP/XS/XA
XP Terminal
XS Balance with “S” Platform
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
12750734 Weighing Platform “S”, XP/XS/XA Each
13552620 NewClassic MS-S 0.01 /0.1 g Balance Each
12780724 XP Terminal Each
13542620 PHS, NewClassic MS 0.1/1 mg Balance Each
11365282 XS Balance with “S” Platform Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 12750734 Supplier Mettler Toledo™ Supplier No. 11133034
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
Protects balances from accidental spills.
  • For weighing platform, S-Platform, XP/XS/XA Precision balances, 10mg/0.1g models
  • Covers platform
  • Material: Barex 210
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Weighing Platform “S”, XP/XS/XA
Material Barex 210
Type Protective Cover