missing translation for 'onlineSavingsMsg'
Learn More

IN-RYD23, TRC

Product Code. 30404086
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30404086 1 mg 1mg
30389109 10 mg 10mg
30404295 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30404086 Supplier Toronto Research Chemicals Supplier No. TRCI4996201MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Bromo-1-(3-chloro-2-pyridinyl)-N-[4-(formylamino)-2-methyl-6-[(methylamino)carbonyl]phenyl]-1H-pyrazole-5-carboxamide
Analyte or Component Names 3-Bromo-1-(3-chloro-2-pyridinyl)-N-[4-(formylamino)-2-methyl-6-[(methylamino)carbonyl]phenyl]-1H-pyrazole-5-carboxamide
Shipping Condition Room Temperature
Molecular Formula C19 H16 Br Cl N6 O3
Synonym IN-RYA13, 3-Bromo-1-(2-chlorophenyl)-N-(4-formamido-2-methyl-6-(methylcarbamoyl)phenyl)-1H-pyrazole-5-carboxamide
InChI Formula InChI=1S/C19H16BrClN6O3/c1-10-6-11(24-9-28)7-12(18(29)22-2)16(10)25-19(30)14-8-15(20)26-27(14)17-13(21)4-3-5-23-17/h3-9H,1-2H3,(H,22,29)(H,24,28)(H,25,30)
Solution Type Neat
SMILES CNC(=O)c1cc(NC=O)cc(C)c1NC(=O)c2cc(Br)nn2c3ncccc3Cl
IUPAC Name 5-bromo-2-(3-chloro-2-pyridyl)-N-[4-formamido-2-methyl-6-(methylcarbamoyl)phenyl]pyrazole-3-carboxamide
Industry Type Pharmaceutical
Molecular Weight (g/mol) 491.72
Quantity 1 mg
Formula Weight 490.0156
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.