missing translation for 'onlineSavingsMsg'
Learn More

IN-Q6S09 (>90%), TRC

Product Code. 30393103
Change view
Click to view available options
Quantity:
0.5 mg
2.5 mg
5 mg
Unit Size:
0.5mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30393103 5 mg 5mg
30397661 2.5 mg 2.5mg
30395656 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30393103 Supplier Toronto Research Chemicals Supplier No. TRCI4996155MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,6,10-trimethyl-5-oxoindazolo[2,3-a]quinazoline-3,8-dicarbonitrile
Analyte or Component Names 1,6,10-trimethyl-5-oxoindazolo[2,3-a]quinazoline-3,8-dicarbonitrile
Shipping Condition Room Temperature
Molecular Formula C19 H13 N5 O
Synonym IN-Q6S09
InChI Formula InChI=1S/C19H13N5O/c1-10-4-12(8-20)6-14-16(10)22-24-17-11(2)5-13(9-21)7-15(17)19(25)23(3)18(14)24/h4-7H,1-3H3
Solution Type Neat
SMILES CN1C(=O)c2cc(cc(C)c2n3nc4c(C)cc(cc4c13)C#N)C#N
IUPAC Name 1,6,10-trimethyl-5-oxoindazolo[2,3-a]quinazoline-3,8-dicarbonitrile
Industry Type Pharmaceutical
Molecular Weight (g/mol) 327.339
Quantity 5 mg
Formula Weight 327.112
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.