missing translation for 'onlineSavingsMsg'
Learn More

IN-KG 433, TRC

Product Code. 30402181
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30402181 250 mg -
30391275 100 mg 100mg
30396771 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30402181 Supplier Toronto Research Chemicals Supplier No. TRCI627995250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 177905-09-8
Molecular Formula C21 H17 Cl F3 N3 O7
Molecular Weight (g/mol) 515.82
Synonym 1H-Indene-2-carboxylic acid, 5-chloro-2,3-dihydro-2-hydroxy-1-[2-[[(methoxycarbonyl)[4-(trifluoromethoxy)phenyl]amino]carbonyl]hydrazinylidene]-, methyl ester, 1H-Indene-2-carboxylic acid, 5-chloro-2,3-dihydro-2-hydroxy-1-[[[(methoxycarbonyl)[4-(trifluoromethoxy)phenyl]amino]carbonyl]hydrazono]-, methyl ester (9CI), IN-KG 433
IUPAC Name methyl (3Z)-6-chloro-2-hydroxy-3-[[methoxycarbonyl-[4-(trifluoromethoxy)phenyl]carbamoyl]hydrazinylidene]-1H-indene-2-carboxylate
SMILES COC(=O)N(C(=O)N\N=C/1\c2ccc(Cl)cc2CC1(O)C(=O)OC)c3ccc(OC(F)(F)F)cc3

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C21H17ClF3N3O7/c1-33-17(29)20(32)10-11-9-12(22)3-8-15(11)16(20)26-27-18(30)28(19(31)34-2)13-4-6-14(7-5-13)35-21(23,24)25/h3-9,32H,10H2,1-2H3,(H,27,30)/b26-16-
Formula Weight 515.0707
Chemical Name or Material Indoxacarb metabolite 2 IN-KG 433
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.