missing translation for 'onlineSavingsMsg'
Learn More

IMR-1A, MedChemExpress

Product Code. 16433018
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16433018 10 mg 10mg
16424326 100 mg 100mg
16419586 50 mg 50mg
16415757 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16433018 Supplier Medchem Express Supplier No. HY100431A
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress IMR-1A, a acid metabolite of IMR-1, is a Notch inhibitor with an IC50 of 0.5 μM. IMR-1A has a 50-fold increase in potency with respect to IMR-1. IMR-1 can metabolize in vivo to IMR-1A.

IMR-1A, a acid metabolite of IMR-1, is a Notch inhibitor with an IC50 of 0.5 μM. IMR-1A has a 50-fold increase in potency with respect to IMR-1. IMR-1 can metabolize in vivo to IMR-1A.

Specifications

Chemical Name or Material IMR-1A
CAS 331862-41-0
Color Yellow
Purity Grade Notes Research
Molecular Formula C13H11NO5S2
Quantity 10 mg
Solubility Information DMSO : 2 mg/mL (6.15 mM; Need ultrasonic) ∣H2O : 1 mg/mL (3.07 mM; ultrasonic and warming and heat to 80°C)
SMILES O=C(O)COC1=CC=C(/C=C(SC(N2)=S)\C2=O)C=C1OC
Molecular Weight (g/mol) 325.36
Formula Weight 325.36
Percent Purity 98.23%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.