Learn More
importin alpha 5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 18137-1-AP-150UL
 View more versions of this product
Product Code. 16848184
  • £349.00 / 150µL
Description
The import of proteins into the nucleus is a process that involves at least 2 steps. The first is an energy-independent docking of the protein to the nuclear envelope and the second is an energy-dependent translocation through the nuclear pore complex. Imported proteins require a nuclear localization sequence (NLS) which generally consists of a short region of basic amino acids or 2 such regions spaced about 10 amino acids apart. Proteins involved in the first step of nuclear import have been identified in different systems. These include the Xenopus protein importin and its yeast homolog, SRP1 (a suppressor of certain temperature sensitive mutations of RNA polymerase I in Saccharomyces cerevisiae), which bind to the NLS. KPNA2 protein interacts with the NLSs of DNA helicase Q1 and SV40 T antigen and may be involved in the nuclear transport of proteins. KPNA2 also may play a role in V(D)J recombination.
Specifications
Specifications
importin alpha 5
Polyclonal
Unconjugated
KPNA1
IMA5, Importin alpha 5, Importin subunit alpha 1, IPOA5, Karyopherin subunit alpha 1, KPNA1, Nucleoprotein interactor 1, RAG cohort protein 2, RCH2, SRP1, SRP1 beta
Rabbit
Antigen Affinity Chromatography
RUO
16646, 288064, 3836
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.17 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P52294, P83953, Q60960
Kpna1
importin alpha5 Fusion Protein Ag12751
150 μL
Primary
Human, Mouse, Rat