missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Importin alpha 5/KPNA1/SRP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-54603
This item is not returnable.
View return policy
Description
Importin alpha 5/KPNA1/SRP1 Polyclonal specifically detects Importin alpha 5/KPNA1/SRP1 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| Importin alpha 5/KPNA1/SRP1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| importin subunit alpha-1, importin-alpha-S1, IPOA5, karyopherin alpha 1 (importin alpha 5), Karyopherin subunit alpha-1, NPI-1SRP1importin alpha 5, Nucleoprotein interactor 1, RCH2RAG cohort protein 2, recombination activating gene cohort 2, SRP1-beta | |
| Rabbit | |
| 60 kDa | |
| 100 μL | |
| Primary | |
| This product is specific to Subunit or Isoform: alpha-1. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
| IgG |
| Western Blot, Immunohistochemistry | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml, Immunohistochemistry | |
| P52294 | |
| KPNA1 | |
| Synthetic peptides corresponding to KPNA1(karyopherin alpha 1 (importin alpha 5)) The peptide sequence was selected from the N terminal of KPNA1. Peptide sequence TTPGKENFRLKSYKNKSLNPDEMRRRREEEGLQLRKQKREEQLFKRRNVA. | |
| Affinity purified | |
| RUO | |
| 3836 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction