Learn More
Importin alpha 5/KPNA1/SRP1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP1-76297-0.025mg
 View more versions of this product
Product Code. 18573412
  • £134.00 / 25µg
Description
Importin alpha 5/KPNA1/SRP1 Polyclonal antibody specifically detects Importin alpha 5/KPNA1/SRP1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Importin alpha 5/KPNA1/SRP1
Polyclonal
Unconjugated
PBS
importin subunit alpha-1, importin-alpha-S1, IPOA5, karyopherin alpha 1 (importin alpha 5), Karyopherin subunit alpha-1, NPI-1SRP1importin alpha 5, Nucleoprotein interactor 1, RCH2RAG cohort protein 2, recombination activating gene cohort 2, SRP1-beta
Antibody was raised against a 15 amino acid synthetic peptide near the amino terminus of human KPNA1. The immunogen is located within amino acids 30 - 80 of KPNA1. Amino Acid Squence: NVATAEEETEEEVMS
0.025 mg
Primary
Human, Mouse, Rat
Purified
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
1 mg/mL
Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/ Immunofluorescence 2.5-20 μg/mL
NP_002255
Rabbit
Peptide affinity purified
RUO
3836
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.