missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Importin alpha 2/KPNA2 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-79807H
This item is not returnable.
View return policy
Description
Importin alpha 2/KPNA2 Polyclonal antibody specifically detects Importin alpha 2/KPNA2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Importin alpha 2/KPNA2 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| importin alpha 1, importin-alpha-P1, IPOA1, karyopherin alpha 2 (RAG cohort 1, importin alpha 1), Karyopherin subunit alpha-2, pendulin, QIP2importin alpha 2, RAG cohort 1, RAG cohort protein 1, RCH1importin subunit alpha-2, SRP1, SRP1alpha, SRP1-alpha | |
| The immunogen recognized by this antibody maps to a region between residues 1 and 50 of human RAG cohort 1 (Karyopherin Alpha 2) using the numbering given in entry NP_002257.1 (GeneID 3838). | |
| 0.1 mL | |
| Cell Cycle and Replication, Cellular Markers, Core ESC Like Genes, Immunology, Stem Cell Markers | |
| 3838 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| HRP | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction