missing translation for 'onlineSavingsMsg'
Learn More

Importin alpha 2/KPNA2 Antibody, DyLight 680, Novus Biologicals™

Product Code. 18409457 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18409457 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18409457 Supplier Novus Biologicals Supplier No. NBP176298FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Importin alpha 2/KPNA2 Polyclonal antibody specifically detects Importin alpha 2/KPNA2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Importin alpha 2/KPNA2
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias importin alpha 1, importin-alpha-P1, IPOA1, karyopherin alpha 2 (RAG cohort 1, importin alpha 1), Karyopherin subunit alpha-2, pendulin, QIP2importin alpha 2, RAG cohort 1, RAG cohort protein 1, RCH1importin subunit alpha-2, SRP1, SRP1alpha, SRP1-alpha
Host Species Rabbit
Immunogen Antibody was raised against a 15 amino acid synthetic peptide near the amino terminus of human KPNA2. The immunogen is located within amino acids 40 - 90 of KPNA2. Amino Acid Squence: SFPDDATSPLQENRN
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Cellular Markers, Core ESC Like Genes, Immunology, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3838
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.