missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Importin 9 Recombinant Rabbit Monoclonal Antibody (23GB5990), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | Importin 9 |
|---|---|
| Clone | 23GB5990 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089088
![]() |
Invitrogen™
MA553936 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089729
|
Invitrogen™
MA553937 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Importin 9 Recombinant Monoclonal antibody specifically detects Importin 9 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| Importin 9 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q91YE6, Q96P70 | |
| 226432, 304817, 55705 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB5990 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 0710008K06Rik; AI845704; C78347; DKFZp761M1547; FLJ10402; HSPC273; Imp9; imp9a; imp9b; importin 9; importin-9; importin-9a; importin-9b; Ipo9; KIAA1192; mKIAA1192; Ran binding protein 9; ran-binding protein 9; RANBP9; RP11-90L20.4 | |
| A synthesized peptide derived from human Importin 9/RANBP9 (91-155AA). | |
| Primary | |
| -20°C | |
| IPO9 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title