missing translation for 'onlineSavingsMsg'
Learn More

Importin-9 Antibody (SR1154), Novus Biologicals™

Rabbit Monoclonal Antibody

£257.00 - £341.00

Specifications

Antigen Importin-9
Clone SR1154
Dilution Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18604315
View Documents
Novus Biologicals
NBP3-22553-100ul
100 μg
£341.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18684315
View Documents
Novus Biologicals
NBP3-22553-50ul
50 μg
£257.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Importin-9 Monoclonal antibody specifically detects Importin-9 in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications

Specifications

Importin-9
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200
Monoclonal
Purified
RUO
Human
DKFZp761M1547, FLJ10402, Imp9IMP9, importin 9, importin-9, KIAA1192, Ran-binding protein 9, RANBP9
A synthesized peptide derived from human Importin-9 (Uniprot #: Q96P70)
Primary
Store at -20° C. Avoid freeze/thaw cycles.
SR1154
Western Blot, Immunohistochemistry
Unconjugated
Rabbit
Core ESC Like Genes, Stem Cell Markers
PBS, pH 7.4, 150mM NaCl, 50% glycerol.
55705
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.