missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Importin-8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NBP2-24751
This item is not returnable.
View return policy
Description
Importin-8 Polyclonal specifically detects Importin-8 in Human, Primate samples. It is validated for Western Blot, Simple Western, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications
| Importin-8 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| FLJ21954, FLJ26580, Imp8, importin 8, importin-8, RAN binding protein 8, Ran-binding protein 8, RanBP8, RANBP8imp8 | |
| Rabbit | |
| Protein G purified | |
| RUO | |
| 10526 | |
| Human, Primate | |
| Purified |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| 1.0 mg/ml | |
| Western Blot 1:200-1:2000, Simple Western 1:20, Immunocytochemistry/Immunofluorescence 1:20-1:2000, Immunoprecipitation reported in scientific literature (PMID 19131589) | |
| O15397 | |
| IPO8 | |
| A portion of amino acids 850-900 of human Importin-8 was used as immunogen for the antibody. | |
| 0.1 mg | |
| Primary | |
| The amino acid sequence used as immunogen for the POU3F2 antibody is 100% homologous in human, horse, pig, mouse, cattle, rat and dog. Alternative initiation produces three different forms of this protein (approximately 47, 29 and 27 kD). | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction