missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Importin 4 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA558634

Product Code. 15746511

  • £467.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: AFLPYMESVF EEVFKLLECP HLNVRKAAHE ALGQFCCALH KACQSCPSEP NTAALQAALA RVVPSYMQAV NRERERQVVM AVLE Highest antigen sequence identity to the following orthologs: Mouse - 69%, Rat - 77%.

Importin-4 is a protein that in humans is encoded by the IPO4 gene. Functions in nuclear protein import as nuclear transport receptor. Serves as receptor for nuclear localization signals (NLS) in cargo substrates. Is thought to mediate docking of the importin/substrate complex to the nuclear pore complex (NPC) through binding to nucleoporin and the complex is subsequently translocated through the pore by an energy requiring, Ran-dependent mechanism. At the nucleoplasmic side of the NPC, Ran binds to the importin, the importin/substrate complex dissociates and importin is re-exported from the nucleus to the cytoplasm where GTP hydrolysis releases Ran.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Importin 4
Polyclonal
Unconjugated
Ipo4
8430408O15Rik; AA409693; Imp4; Imp4a; IMP4B; importin 4; importin 4a; importin-4; Importin-4a; importin-4b; IPO4; Ran-binding protein 4; ranBP4
Rabbit
Antigen affinity chromatography
RUO
79711
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.4 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q8TEX9
Ipo4
Recombinant protein corresponding to Human Importin 4. Recombinant protein control fragment (Product #RP-97613).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.