missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Importin 13 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA521884
This item is not returnable.
View return policy
Description
Recommended positive controls: HeLa, Importin 13 tansfected 293T cell. Predicted reactivity: Mouse (100%), Rat (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a member of the importin-beta family of nuclear transport proteins. The encoded protein mediates the import of specific cargo proteins from the cytoplasm to the nucleus and is dependent on the Ras-related nuclear protein-GTPase system. The encoded protein is also involved in nuclear export of the eukaryotic translation initiation factor 1A.
Specifications
| Importin 13 | |
| Polyclonal | |
| Unconjugated | |
| IPO13 | |
| Imp13; importin 13; importin-13; Ipo13; Kap13; karyopherin 13; karyopherin-13; KIAA0724; late gestation lung 2; late gestation lung 2 protein; LGL2; Ran binding protein 13; ran-binding protein 13; RanBP13; RP11-7O11.4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 116458, 9670 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 1.02 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| O94829, Q9JM04 | |
| IPO13 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human Importin 13. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction