missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Importin 11 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-03801-100UL
This item is not returnable.
View return policy
Description
Importin 11 Polyclonal specifically detects Importin 11 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| Importin 11 | |
| Polyclonal | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 51194 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| imp11, importin 11, Ran binding protein 11, Ran-binding protein 11, RANBP11, RanBP11importin-11, SLRN | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 776-975 of human Importin 11 (NP_057422.3). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction